1-(Difluoromethyl)-3-fluoro-5-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

071573-1G
102520-362EA 412.31 USD
102520-362
1-(Difluoromethyl)-3-fluoro-5-(trifluoromethyl)benzene ≥95%
1-(Difluoromethyl)-3-fluoro-5-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₄F₆
MDL Number: MFCD14698651
CAS Number: 1161362-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR