2-(Difluoromethyl)-1-fluoro-4-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

071572-1G
102520-360EA 235.38 USD
102520-360
2-(Difluoromethyl)-1-fluoro-4-(trifluoromethyl)benzene ≥95%
2-(Difluoromethyl)-1-fluoro-4-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₄F₆
MDL Number: MFCD14698647
CAS Number: 1214334-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR