1-(Difluoromethyl)-2-fluoro-4-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

071570-1G
102520-356EA 287.69 USD
102520-356
1-(Difluoromethyl)-2-fluoro-4-(trifluoromethyl)benzene ≥95%
1-(Difluoromethyl)-2-fluoro-4-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₄F₆
MDL Number: MFCD14698646
CAS Number: 1214364-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR