2-(Difluoromethyl)quinoline ≥97%

Supplier: Matrix Scientific

071538-1G
102520-294EA 423.08 USD
102520-294
2-(Difluoromethyl)quinoline ≥97%
2-(Difluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₇F₂N
MDL Number: MFCD11226558
CAS Number: 1075184-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR