4-Isopropylbenzotrifluoride ≥95%

Supplier: Matrix Scientific

071534-1G
102520-286EA 223.08 USD
102520-286
4-Isopropylbenzotrifluoride ≥95%
4-Isopropylbenzotrifluoride

Hazard: Irritant

Formula: C₁₀H₁₁F₃
MDL Number: MFCD00190107
CAS Number: 32445-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR