(1,1-Difluoropropyl)benzene ≥95%

Supplier: Matrix Scientific

071531-1G
102520-280EA 235.38 USD
102520-280
(1,1-Difluoropropyl)benzene ≥95%
(1,1-Difluoropropyl)benzene

Hazard: Irritant

Formula: C₉H₁₀F₂
Boiling Pt: 25 °C (2 mmHg)
MDL Number: MFCD14525542
CAS Number: 74185-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR