1-(Difluoromethyl)-2,3,4,5,6-pentafluorobenzene ≥97%

Supplier: Matrix Scientific

071524-1G
102520-266EA 235.38 USD
102520-266
1-(Difluoromethyl)-2,3,4,5,6-pentafluorobenzene ≥97%
1-(Difluoromethyl)-2,3,4,5,6-pentafluorobenzene

Hazard: Irritant

Formula: C₇HF₇
MDL Number: MFCD00032556
CAS Number: 22006-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR