3-(Trifluoromethyl)quinoline ≥97%

Supplier: Matrix Scientific

071517-1G
102520-254EA 396.92 USD
102520-254
3-(Trifluoromethyl)quinoline ≥97%
3-(Trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₆F₃N
MDL Number: MFCD11007731
CAS Number: 25199-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR