4-(1,1-Difluoroethyl)pyridine ≥95%

Supplier: Matrix Scientific

071514-1G
102520-248EA 267.69 USD
102520-248
4-(1,1-Difluoroethyl)pyridine ≥95%
4-(1,1-Difluoroethyl)pyridine

Hazard: Irritant

Formula: C₇H₇F₂N
MDL Number: MFCD12404950
CAS Number: 114490-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR