(2,2-Difluoro-1-methylethyl)benzene ≥97%

Supplier: Matrix Scientific

071492-1G
102520-206EA 241.54 USD
102520-206
(2,2-Difluoro-1-methylethyl)benzene ≥97%
(2,2-Difluoro-1-methylethyl)benzene

Hazard: Irritant

Formula: C₉H₁₀F₂
MDL Number: MFCD11226606
CAS Number: 57514-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR