1-Nitro-2,4-bis-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Bis(trifluoromethyl)nitrobenzene

071485-1G
102520-192EA 446.15 USD
102520-192
1-Nitro-2,4-bis-(trifluoromethyl)benzene ≥95%
1-Nitro-2,4-bis-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₃F₆NO₂
MW: 259.11 g/mol
MDL Number: MFCD13185347
CAS Number: 224044-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR