1-Phenoxy-3-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

071456-1G
102520-126EA 272.31 USD
102520-126
1-Phenoxy-3-(trifluoromethyl)benzene ≥95%
1-Phenoxy-3-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₁₃H₉F₃O
MDL Number: MFCD12032175
CAS Number: 330-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR