2,3-Bis(trifluoromethyl)pyrazine ≥95%

Supplier: Matrix Scientific

071455-1G
102520-124EA 270.77 USD
102520-124
2,3-Bis(trifluoromethyl)pyrazine ≥95%
2,3-Bis(trifluoromethyl)pyrazine

Hazard: Irritant

Formula: C₆H₂F₆N₂
MDL Number: MFCD11226566
CAS Number: 1186195-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR