6-Chloro-7-methyl-7H-purine ≥97%

Supplier: Matrix Scientific

071425-500MG
102520-020EA 2396.92 USD
102520-020
6-Chloro-7-methyl-7H-purine ≥97%
6-Chloro-7-methyl-7H-purine

Hazard: Irritant

Formula: C₆H₅ClN₄
MDL Number: MFCD01943043
CAS Number: 5440-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR