2-Chloro-5-(difluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

071387-1G
102519-970EA 173.85 USD
102519-970
2-Chloro-5-(difluoromethyl)pyridine ≥97%
2-Chloro-5-(difluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄ClF₂N
Boiling Pt: 54 °C (4 mmHg)
Density: 1.368
Flash Pt: 107 °C
MDL Number: MFCD16140199
CAS Number: 71701-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR