[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Isopropylisoxazol-5-amine
071375-1G
071375-500MG
102519-946EA
504.62
USD
102519-946
102519-944
[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine ≥97%
[3-(Propan-2-yl)-1,2-oxazol-5-yl]amine
Hazard: Irritant
Formula:
C₆H₁₀N₂O MW: 126.16 g/mol Melting Pt: 67…69 °C |
MDL Number:
MFCD06335179 CAS Number: 88786-11-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...