4-[3-Nitro-4-(piperidin-1-yl)phenyl]-1,2-dihydrophthalazin-1-one ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(3-Nitro-4-piperidin-1-ylphenyl)phthalazin-1(2H)-one

071371-1G 071371-500MG
102519-932EA 504.62 USD
102519-932 102519-930
4-[3-Nitro-4-(piperidin-1-yl)phenyl]-1,2-dihydrophthalazin-1-one ≥97%
4-[3-Nitro-4-(piperidin-1-yl)phenyl]-1,2-dihydrophthalazin-1-one

Hazard: Irritant

Formula: C₁₉H₁₈N₄O₃
MW: 350.38 g/mol
MDL Number: MFCD00116203
CAS Number: 218144-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR