3-Methyl-1,3-oxazolidine-2,4-dione ≥97%
Supplier: Matrix Scientific
071365-1G
071365-500MG
102519-908EA
504.62
USD
102519-908
102519-906
3-Methyl-1,3-oxazolidine-2,4-dione ≥97%
3-Methyl-1,3-oxazolidine-2,4-dione
Hazard: Irritant
Formula:
C₄H₅NO₃ |
MDL Number:
MFCD22126130 CAS Number: 14422-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...