Methyl-3-methyl-1-benzofuran-2-carboxylate ≥97%

Supplier: Matrix Scientific

071362-500MG 071362-1G
102519-896EA 504.62 USD
102519-896 102519-894
Methyl-3-methyl-1-benzofuran-2-carboxylate ≥97%
Methyl-3-methyl-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
MDL Number: MFCD09270117
CAS Number: 2076-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR