Methyl-6-bromo-1-benzofuran-2-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-bromobenzofuran-2-carboxylate
071355-1G
071355-500MG
102519-868EA
504.62
USD
102519-868
102519-866
Methyl-6-bromo-1-benzofuran-2-carboxylate ≥97%
Methyl-6-bromo-1-benzofuran-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇BrO₃ MW: 255.07 g/mol |
MDL Number:
MFCD22126097 CAS Number: 425675-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...