3-N-Cyclohexylpyridine-2,3-diamine ≥97%
Supplier: Matrix Scientific
071337-500MG
071337-1G
071337-5G
102519-792EA
566.15
USD
102519-792
102519-790
102519-788
3-N-Cyclohexylpyridine-2,3-diamine ≥97%
3-N-Cyclohexylpyridine-2,3-diamine
Hazard: Irritant
Formula:
C₁₁H₁₇N₃ MW: 191.28 g/mol Melting Pt: 183…187 °C |
MDL Number:
MFCD20688652 CAS Number: 1286273-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...