6-Bromo-2-fluoropyridin-3-amine ≥97%
Supplier: Matrix Scientific
071326-1G
071326-500MG
102519-746EA
749.23
USD
102519-746
102519-744
6-Bromo-2-fluoropyridin-3-amine ≥97%
6-Bromo-2-fluoropyridin-3-amine
Hazard: Irritant
Formula:
C₅H₄BrFN₂ Melting Pt: 96…97 °C |
MDL Number:
MFCD13659314 CAS Number: 850220-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...