3-(Aminomethyl)-N-methylbenzamide ≥97%

Supplier: Matrix Scientific

071183-5G
102519-582EA 1592.31 USD
102519-582
3-(Aminomethyl)-N-methylbenzamide ≥97%
3-(Aminomethyl)-N-methylbenzamide

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD09048986
CAS Number: 515131-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR