1,3-Dibromo-2-(bromomethyl)benzene ≥97%

Supplier: Matrix Scientific

071180-5G
102519-578EA 1592.31 USD
102519-578
1,3-Dibromo-2-(bromomethyl)benzene ≥97%
1,3-Dibromo-2-(bromomethyl)benzene

Hazard: Irritant

Formula: C₇H₅Br₃
MDL Number: MFCD13185505
CAS Number: 93701-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR