Methyl-2,4-dicyanobenzoate ≥97%

Supplier: Matrix Scientific

071178-5G
102519-572EA 1592.31 USD
102519-572
Methyl-2,4-dicyanobenzoate ≥97%
Methyl-2,4-dicyanobenzoate

Hazard: Irritant

Formula: C₁₀H₆N₂O₂
MDL Number: MFCD22199275
CAS Number: 58331-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR