4-Bromo-5-methylbenzene-1,2-diamine ≥97%

Supplier: Matrix Scientific

071174-5G
102519-564EA 530.77 USD
102519-564
4-Bromo-5-methylbenzene-1,2-diamine ≥97%
4-Bromo-5-methylbenzene-1,2-diamine

Hazard: Irritant

Formula: C₇H₉BrN₂
MDL Number: MFCD16987757
CAS Number: 102169-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR