(1-Ethyl-1H-1,2,4-triazol-5-yl)methanol ≥97%

Supplier: Matrix Scientific

071165-5G
102519-550EA 1592.31 USD
102519-550
(1-Ethyl-1H-1,2,4-triazol-5-yl)methanol ≥97%
(1-Ethyl-1H-1,2,4-triazol-5-yl)methanol

Hazard: Irritant

Formula: C₅H₉N₃O
Melting Pt: 82…84 °C
MDL Number: MFCD09966168
CAS Number: 215868-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR