3,6-Dichloropyrazine-2-carbonitrile ≥97%

Supplier: Matrix Scientific

071159-5G
102519-546EA 2350.77 USD
102519-546
3,6-Dichloropyrazine-2-carbonitrile ≥97%
3,6-Dichloropyrazine-2-carbonitrile

Hazard: Irritant

Formula: C₅HCl₂N₃
MDL Number: MFCD22199269
CAS Number: 356783-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR