3-(4-Nitro-1H-imidazol-1-yl)cyclobutanone ≥97%

Supplier: Matrix Scientific

071140-5G
102519-528EA 1592.31 USD
102519-528
3-(4-Nitro-1H-imidazol-1-yl)cyclobutanone ≥97%
3-(4-Nitro-1H-imidazol-1-yl)cyclobutanone

Hazard: Irritant

Formula: C₇H₇N₃O₃
MDL Number: MFCD22199262
CAS Number: 716316-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR