Ethyl-3,5-diamino-1H-pyrazole-4-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3,5-diamino-1H-pyrazole-4-carboxylate

071136-5G
102519-524EA 1615.38 USD
102519-524
Ethyl-3,5-diamino-1H-pyrazole-4-carboxylate ≥97%
Ethyl-3,5-diamino-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₆H₁₀N₄O₂
MW: 170.17 g/mol
MDL Number: MFCD04070845
CAS Number: 6825-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR