5-Isopropoxy-1H-pyrazol-3-amine ≥97%

Supplier: Matrix Scientific

071128-5G
102519-514EA 2784.62 USD
102519-514
5-Isopropoxy-1H-pyrazol-3-amine ≥97%
5-Isopropoxy-1H-pyrazol-3-amine

Hazard: Irritant

Formula: C₆H₁₁N₃O
MDL Number: MFCD11846994
CAS Number: 121507-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR