2-Methyl-6-[bis(tert-butoxycarbonyl)amino]-4-chloropyrimidine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-[Bis(Boc)amino]-4-chloro-2-methylpyrimidine

071124-5G
102519-508EA 1592.31 USD
102519-508
2-Methyl-6-[bis(tert-butoxycarbonyl)amino]-4-chloropyrimidine ≥97%
2-Methyl-6-[bis(tert-butoxycarbonyl)amino]-4-chloropyrimidine

Hazard: Irritant

Formula: C₁₅H₂₂ClN₃O₄
MW: 343.81 g/mol
MDL Number: MFCD22199258
CAS Number: 1350918-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR