(4-Amino-2-(methylthio)pyrimidin-5-yl)methanol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-5-hydroxymethyl-2-(methylthio)pyrimidine

071115-5G
102519-496EA 1592.31 USD
102519-496
(4-Amino-2-(methylthio)pyrimidin-5-yl)methanol ≥97%
(4-Amino-2-(methylthio)pyrimidin-5-yl)methanol

Hazard: Irritant

Formula: C₆H₉N₃OS
MW: 171.22 g/mol
MDL Number: MFCD08236793
CAS Number: 588-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR