2-Amino-5-butyl-4-methyl-1H-pyrrole-3-carbonitrile ≥97%

Supplier: Matrix Scientific

071100-5G
102519-466EA 1592.31 USD
102519-466
2-Amino-5-butyl-4-methyl-1H-pyrrole-3-carbonitrile ≥97%
2-Amino-5-butyl-4-methyl-1H-pyrrole-3-carbonitrile

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MDL Number: MFCD22199253
CAS Number: 1227958-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR