5-Bromo-6-methoxynicotinaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-methoxypyridine-5-carboxyaldehyde

071093-5G
102519-450EA 2766.15 USD
102519-450
5-Bromo-6-methoxynicotinaldehyde ≥97%
5-Bromo-6-methoxynicotinaldehyde

Hazard: Irritant/Store in Freezer

Formula: C₇H₆BrNO₂
MW: 216.04 g/mol
MDL Number: MFCD16606522
CAS Number: 65873-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR