3-(tert-Butoxycarbonylamino)-2-methoxyisonicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3-((tert-Butoxycarbonyl)amino)-2-methoxyisonicotinic acid

071077-5G
102519-436EA 1592.31 USD
102519-436
3-(tert-Butoxycarbonylamino)-2-methoxyisonicotinic acid ≥97%
3-(tert-Butoxycarbonylamino)-2-methoxyisonicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₅
MW: 268.27 g/mol
MDL Number: MFCD22199249
CAS Number: 870997-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR