(2R,3R)-Diethyl-2-bromo-3-hydroxysuccinate ≥97%

Supplier: Matrix Scientific

071282-5G
102519-676EA 1592.31 USD
102519-676
(2R,3R)-Diethyl-2-bromo-3-hydroxysuccinate ≥97%
(2R,3R)-Diethyl-2-bromo-3-hydroxysuccinate

Hazard: Irritant

Formula: C₈H₁₃BrO₅
MDL Number: MFCD21648267
CAS Number: 80640-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR