1-Isoquinolinol ≥97%
Supplier: Matrix Scientific
Synonyms:
Isocarbostyril
, 1-Hydroxyisoquinoline
071269-5G
102519-650EA
181.54
USD
102519-650
1-Isoquinolinol ≥97%
1-Isoquinolinol
Hazard: Irritant
Formula:
C₉H₇NO MW: 145.16 g/mol Melting Pt: 212…215 °C |
MDL Number:
MFCD00006899 CAS Number: 491-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...