4-Chloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥97%
Supplier: Matrix Scientific
071253-5G
071253-1G
102551-780EA
396.92
USD
102551-780
102519-646
4-Chloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥97%
4-Chloro-5-methyl-7H-pyrrolo[2,3-d]pyrimidine
Hazard: Irritant
Formula:
C₇H₆ClN₃ |
MDL Number:
MFCD09264586 CAS Number: 1618-36-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...