2-Chlorothieno[3,2-d]pyrimidine-4-carbonitrile ≥97%

Supplier: Matrix Scientific

071247-5G
102519-642EA 1592.31 USD
102519-642
2-Chlorothieno[3,2-d]pyrimidine-4-carbonitrile ≥97%
2-Chlorothieno[3,2-d]pyrimidine-4-carbonitrile

Hazard: Irritant

Formula: C₇H₂ClN₃S
MDL Number: MFCD21648256
CAS Number: 1364663-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR