1-(2-Methoxy-6-(4-methoxybenzyloxy)phenyl)ethanone ≥97%

Supplier: Matrix Scientific

Synonyms: 2'-Methoxy-6'-(4-methoxybenzyloxy)acetophenone

071213-5G
102519-616EA 1592.31 USD
102519-616
1-(2-Methoxy-6-(4-methoxybenzyloxy)phenyl)ethanone ≥97%
1-(2-Methoxy-6-(4-methoxybenzyloxy)phenyl)ethanone

Hazard: Irritant

Formula: C₁₇H₁₈O₄
MW: 286.33 g/mol
MDL Number: MFCD21648248
CAS Number: 1234015-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR