(R)-Methyl-4-(3,4-dimethylpiperazin-1-yl)benzoate ≥97%

Supplier: Matrix Scientific

071212-5G
102519-614EA 1592.31 USD
102519-614
(R)-Methyl-4-(3,4-dimethylpiperazin-1-yl)benzoate ≥97%
(R)-Methyl-4-(3,4-dimethylpiperazin-1-yl)benzoate

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₂
MDL Number: MFCD21648247
CAS Number: 1201670-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR