2-Methoxy-4-(4-methylpiperazin-1-yl)aniline ≥97%

Supplier: Matrix Scientific

071210-5G
102519-610EA 730.77 USD
102519-610
2-Methoxy-4-(4-methylpiperazin-1-yl)aniline ≥97%
2-Methoxy-4-(4-methylpiperazin-1-yl)aniline

Hazard: Irritant

Formula: C₁₂H₁₉N₃O
MW: 221.2988 g/mol
MDL Number: MFCD11848954
CAS Number: 122833-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR