Benzyl-2,4-difluoro-3-formylphenylcarbamate ≥97%

Supplier: Matrix Scientific

071204-5G
102519-602EA 1592.31 USD
102519-602
Benzyl-2,4-difluoro-3-formylphenylcarbamate ≥97%
Benzyl-2,4-difluoro-3-formylphenylcarbamate

Hazard: Irritant

Formula: C₁₅H₁₁F₂NO₃
MW: 291.26 g/mol
MDL Number: MFCD21648244
CAS Number: 918524-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR