7-Fluoro-1H-indazole-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

071228-5G
102519-636EA 900 USD
102519-636
7-Fluoro-1H-indazole-3-carboxylic acid ≥97%
7-Fluoro-1H-indazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₅FN₂O₂
MW: 180.14 g/mol
Boiling Pt: 445 °C
Density: 1.611
Flash Pt: 223 °C
MDL Number: MFCD07378920
CAS Number: 959236-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR