4-Indoleboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (1H-Indol-4-yl)boronic acid4-Indolylboronic acid

071223-5G
102519-632EA 946.15 USD
102519-632
4-Indoleboronic acid ≥97%
4-Indoleboronic acid

Hazard: Irritant

Formula: C₈H₈BNO₂
MW: 160.97 g/mol
Boiling Pt: 433 °C
Density: 1.33
Flash Pt: 215 °C
MDL Number: MFCD03095175
CAS Number: 220465-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR