(R)-Methyl-4-(3-methylpiperazin-1-yl)benzoate ≥97%

Supplier: Matrix Scientific

071211-5G
102519-612EA 946.15 USD
102519-612
(R)-Methyl-4-(3-methylpiperazin-1-yl)benzoate ≥97%
(R)-Methyl-4-(3-methylpiperazin-1-yl)benzoate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD11112309
CAS Number: 1201670-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR