1-(3-Methoxy-4-nitrophenyl)-4-methylpiperazine ≥97%

Supplier: Matrix Scientific

071209-5G
102519-608EA 946.15 USD
102519-608
1-(3-Methoxy-4-nitrophenyl)-4-methylpiperazine ≥97%
1-(3-Methoxy-4-nitrophenyl)-4-methylpiperazine

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃
MDL Number: MFCD16249532
CAS Number: 761440-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR