N-Methyl-O-toluamide ≥97%

Supplier: Matrix Scientific

071208-5G
102519-606EA 946.15 USD
102519-606
N-Methyl-O-toluamide ≥97%
N-Methyl-o-toluamide

Hazard: Irritant

Formula: C₈H₉NO
Boiling Pt: 278 °C
Melting Pt: 73…77 °C
Density: 1.021
Flash Pt: 158 °C
CAS Number: 2170-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR