Benzyl-2,4-difluorophenylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: Benzyl (2,4-difluorophenyl)carbamate

071203-5G
102519-600EA 946.15 USD
102519-600
Benzyl-2,4-difluorophenylcarbamate ≥97%
Benzyl-2,4-difluorophenylcarbamate

Hazard: Irritant

Formula: C₁₄H₁₁F₂NO₂
MW: 263.25 g/mol
MDL Number: MFCD21648243
CAS Number: 112434-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR